siRNA Details
| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 4 | 1334 | ||
| GenBank mRNA | AK090377.1 | - | 4 | 1335 | ||
| AK157715.1 | - | 4 | 1283 | |||
| X85788.1 | - | 1 | 884 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dcc| Gene Symbol: Dcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007831.3 | NP_031857.2 | 4 | 1334 | ||
| GenBank mRNA | AK090377.1 | - | 4 | 1335 | ||
| AK157715.1 | - | 4 | 1283 | |||
| X85788.1 | - | 1 | 884 | |||