siRNA Details
Gene Symbol: Asap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276461.1 | NP_001263390.1 | 3 | 349 | ||
NM_001276462.1 | NP_001263391.1 | 1 | 258 | |||
NM_001276463.1 | NP_001263392.1 | 1 | 258 | |||
NM_001276467.1 | NP_001263396.1 | 3 | 349 | |||
NM_010026.3 | NP_034156.2 | 3 | 349 | |||
GenBank mRNA | AF075461.1 | - | 3 | 168 | ||
AF075462.1 | - | 3 | 168 | |||
AK132979.1 | - | 1 | 88 | |||
AK146545.1 | - | 3 | 187 | |||
AK147689.1 | - | 1 | 259 | |||
AK161586.1 | - | 259 | ||||
BC048818.1 | - | 1 | 224 | |||
BC094581.1 | - | 3 | 349 | 30549241 | ||
CB527210.1 | - | 1 | 52 | |||
CF735731.1 | - | 3 | 349 |
