siRNA Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290183.1 | NP_001277112.1 | 3 | 364 | ||
| NM_007837.4 | NP_031863.3 | 4 | 412 | |||
| GenBank mRNA | AK028120.1 | - | 4 | 409 | ||
| BC013718.1 | - | 4 | 407 | 4012433 | ||
| BE134148.1 | - | 1 | 464 | |||
| CF982562.1 | - | 1 | 62 | |||
| X67083.1 | - | 3 | 319 | |||

Ddit3
Genomic Map