siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007838.2 | NP_031864.2 | 4 | 532 | ||
| GenBank mRNA | AK077651.1 | - | 4 | 395 | ||
| AK081542.1 | - | 4 | 424 | |||
| AK084267.1 | - | 4 | 416 | |||
| AK150683.1 | - | 4 | 423 | |||
| AK182353.1 | - | 2 | 68 | |||
| AK200087.1 | - | 2 | 93 | |||
| BC068132.1 | - | 4 | 532 | 6808198 | ||
| D89063.1 | - | 4 | 534 | |||

Ddost
Genomic Map