siRNA Details
| Gene Symbol: Dhx9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007842.2 | NP_031868.2 | 11 | 1236 | ||
| GenBank mRNA | AK141824.1 | - | 11 | 1231 | ||
| AK148423.1 | - | 11 | 1232 | |||
| AY512925.1 | - | 7 | 692 | |||
| BC089159.1 | - | 11 | 1230 | |||
| U91922.1 | - | 11 | 1193 | |||

Dhx9
Genomic Map