siRNA Details
| Gene Symbol: Twist2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007855.2 | NP_031881.1 | 1 | 251 | ||
| GenBank mRNA | AK011180.1 | - | 1 | 267 | ||
| AK011861.1 | - | 1 | 268 | |||
| BC090636.1 | - | 1 | 256 | 30942245 | ||
| U36384.1 | - | 1 | 251 | |||

Twist2
Genomic Map