siRNA Details
| Gene Symbol: Dgcr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289813.1 | NP_001276742.1 | 4 | 506 | ||
| NM_001289814.1 | NP_001276743.1 | 4 | 497 | |||
| NM_010047.4 | NP_034177.1 | 5 | 694 | |||
| GenBank mRNA | AF021031.1 | - | 4 | 398 | ||
| AK019346.1 | - | 5 | 697 | |||
| AK160342.1 | - | 5 | 684 | |||
| BC048503.1 | - | 4 | 349 | 6770478 | ||
| BE945982.1 | - | 4 | 406 | |||

Dgcr6
Genomic Map