siRNA Details
| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153150.2 | NP_694790.1 | 7 | 839 | ||
| GenBank mRNA | AK005070.1 | - | 7 | 769 | ||
| AK159289.1 | - | 7 | 802 | |||
| AK170191.1 | - | 7 | 803 | |||
| BC037087.1 | - | 7 | 799 | 5124403 | ||

Slc25a1
Genomic Map