siRNA Details
| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007857.4 | NP_031883.1 | 1 | 388 | ||
| GenBank mRNA | AK032466.1 | - | 1 | 371 | ||
| AK033392.1 | - | 1 | 389 | |||
| BC062810.1 | - | 1 | 354 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhh| Gene Symbol: Dhh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007857.4 | NP_031883.1 | 1 | 388 | ||
| GenBank mRNA | AK032466.1 | - | 1 | 371 | ||
| AK033392.1 | - | 1 | 389 | |||
| BC062810.1 | - | 1 | 354 | |||