siRNA Details
| Gene Symbol: Dffb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007859.4 | NP_031885.3 | 2 | 302 | ||
| GenBank mRNA | AB009377.1 | - | 2 | 210 | ||
| AK036094.1 | - | 2 | 279 | |||
| AK077619.1 | - | 2 | 348 | |||
| AK089267.1 | - | 2 | 279 | |||
| BC053052.1 | - | 2 | 232 | 6403143 | ||

Dffb
Genomic Map