siRNA Details
| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007868.5 | NP_031894.1 | 13 | 1840 | ||
| GenBank mRNA | AK044536.1 | - | 13 | 1640 | ||
| M18025.1 | - | 5 | 698 | |||
| M68859.1 | - | 13 | 1813 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmd| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007868.5 | NP_031894.1 | 13 | 1840 | ||
| GenBank mRNA | AK044536.1 | - | 13 | 1640 | ||
| M18025.1 | - | 5 | 698 | |||
| M68859.1 | - | 13 | 1813 | |||