siRNA Details
| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007868.5 | NP_031894.1 | 23 | 3308 | ||
| GenBank mRNA | M18025.1 | - | 15 | 2166 | ||
| M68859.1 | - | 23 | 3281 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmd| Gene Symbol: Dmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007868.5 | NP_031894.1 | 23 | 3308 | ||
| GenBank mRNA | M18025.1 | - | 15 | 2166 | ||
| M68859.1 | - | 23 | 3281 | |||