siRNA Details
| Gene Symbol: Dnah8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013811.3 | NP_038839.2 | 13258 | |||
| GenBank mRNA | AF356520.1 | - | 13258 | |||
| AF356522.1 | - | 13268 | ||||
| AK133238.1 | - | 14 | 2013 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah8| Gene Symbol: Dnah8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013811.3 | NP_038839.2 | 13258 | |||
| GenBank mRNA | AF356520.1 | - | 13258 | |||
| AF356522.1 | - | 13268 | ||||
| AK133238.1 | - | 14 | 2013 | |||