siRNA Details
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190817.1 | NP_001177746.1 | 1, 2 | 541 | ||
NM_007869.3 | NP_031895.1 | 1, 2 | 541 | |||
GenBank mRNA | AK012505.1 | - | 1, 2 | 535 | ||
AK133582.1 | - | 1, 2 | 525 | |||
AK144228.1 | - | 1, 2 | 497 | |||
BC020071.1 | - | 426 | ||||
BC080300.1 | - | 1, 2 | 522 | |||
L16953.1 | - | 1, 2 | 218 |
