siRNA Details
| Gene Symbol: Dnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039520.2 | NP_001034609.1 | 19 | 2275 | ||
| NM_001253893.1 | NP_001240822.1 | 19 | 2275 | |||
| NM_001253894.1 | NP_001240823.1 | 19 | 2275 | |||
| NM_007871.2 | NP_031897.2 | 18 | 2263 | |||
| GenBank mRNA | AK005012.2 | - | 19 | 2251 | ||
| AK150046.1 | - | 19 | 2237 | |||
| AK154375.1 | - | 17 | 1926 | |||
| AK157844.1 | - | 19 | 2244 | |||
| AK170574.1 | - | 19 | 2251 | |||
| AK171049.1 | - | 19 | 2229 | |||
| AK171566.1 | - | 19 | 2240 | |||
| AK172231.1 | - | 19 | 2256 | |||
| AK193376.1 | - | 2 | 99 | |||
| AK199747.1 | - | 2 | 99 | |||
| BC010233.1 | - | 10 | 991 | 3499181 | ||
| L31398.1 | - | 18 | 2203 | |||

Dnm2
Genomic Map