siRNA Details
| Gene Symbol: Dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010073.2 | NP_034203.1 | 4 | 262 | ||
| GenBank mRNA | AB013360.1 | - | 4 | 234 | ||
| AK002468.1 | - | 4 | 246 | |||
| AK004241.1 | - | 4 | 237 | |||
| BC008256.1 | - | 4 | 262 | 2648752 | ||

Dpm2
Genomic Map