siRNA Details
Gene Symbol: Drg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021354.1 | NP_067329.1 | 8 | 779 | ||
GenBank mRNA | AJ243590.1 | - | 8 | 779 | ||
AK012988.1 | - | 8 | 747 | |||
BC082564.1 | - | 8 | 735 | 30551418 |
