siRNA Details
| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291809.1 | NP_001278738.1 | 13 | 2147 | ||
| NM_007882.3 | NP_031908.3 | 13 | 2147 | |||
| GenBank mRNA | BC144845.1 | - | 13 | 2132 | ||
| Y11169.1 | - | 13 | 2147 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dsc3| Gene Symbol: Dsc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291809.1 | NP_001278738.1 | 13 | 2147 | ||
| NM_007882.3 | NP_031908.3 | 13 | 2147 | |||
| GenBank mRNA | BC144845.1 | - | 13 | 2132 | ||
| Y11169.1 | - | 13 | 2147 | |||