siRNA Details
Gene Symbol: Epyc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007884.2 | NP_031910.1 | 8 | 1006 | ||
GenBank mRNA | AK005684.1 | - | 8 | 1008 | ||
AK015223.1 | - | 8 | 1006 | |||
AK137179.1 | - | 7 | 931 | |||
AK157900.1 | - | 1257 | ||||
AK157901.1 | - | 7 | 1268 | |||
BC012695.1 | - | 7 | 978 | 4037028 | ||
D78274.1 | - | 7 | 921 |
