siRNA Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF126967.1 | - | 4 | 516 | ||
| AK033285.1 | - | 4 | 525 | |||
| AK077014.1 | - | 574 | ||||
| AK144176.1 | - | 4 | 470 | |||
| BC005634.1 | - | 4 | 445 | |||
| BC031757.1 | - | 4 | 499 | |||
| BC046459.1 | - | 4 | 583 | 5322519 | ||
| BC057011.1 | - | 3 | 269 | |||
| BC071228.1 | - | 4 | 526 | 4915902 | ||
| X76858.1 | - | 3 | 309 | |||

E4f1
Genomic Map