siRNA Details
| Gene Symbol: Ebf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290709.1 | NP_001277638.1 | 14 | 1540 | ||
| NM_001290711.1 | NP_001277640.1 | 14 | 1656 | |||
| NM_007897.3 | NP_031923.1 | 14 | 1537 | |||
| GenBank mRNA | AK031239.1 | - | 14 | 1535 | ||
| AK035781.1 | - | 14 | 1420 | |||
| AK137389.1 | - | 14 | 1578 | |||
| AK141175.1 | - | 14 | 1540 | |||
| BC139362.1 | - | 14 | 1498 | 8862384 | ||
| L12147.1 | - | 14 | 1503 | |||

Ebf1
Genomic Map