siRNA Details
| Gene Symbol: Ecel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277925.1 | NP_001264854.1 | 6, 7 | 1401 | ||
| NM_021306.3 | NP_067281.2 | 6, 7 | 1393 | |||
| GenBank mRNA | AB026294.1 | - | 5, 6 | 1221 | ||
| AK039467.1 | - | 6, 7 | 1394 | |||
| AK159365.1 | - | 6, 7 | 1403 | |||
| BC057569.1 | - | 5, 6 | 1223 | 6413731 | ||

Ecel1
Genomic Map