siRNA Details
| Gene Symbol: Edn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007902.2 | NP_031928.2 | 5 | 543 | ||
| GenBank mRNA | BC037042.1 | - | 5 | 772 | 4512195 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edn2| Gene Symbol: Edn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007902.2 | NP_031928.2 | 5 | 543 | ||
| GenBank mRNA | BC037042.1 | - | 5 | 772 | 4512195 | |