siRNA Details
Gene Symbol: Efna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007910.2 | NP_031936.2 | 2 | 371 | ||
GenBank mRNA | AK012195.1 | - | 2 | 375 | ||
AK142604.1 | - | 2 | 372 | |||
U90663.1 | - | 2 | 371 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007910.2 | NP_031936.2 | 2 | 371 | ||
GenBank mRNA | AK012195.1 | - | 2 | 375 | ||
AK142604.1 | - | 2 | 372 | |||
U90663.1 | - | 2 | 371 |