siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 4, 5 | 555 | ||
| GenBank mRNA | AF035527.1 | - | 3, 4 | 553 | ||
| AK028651.1 | - | 4, 5 | 526 | |||
| BC005520.1 | - | 5, 6 | 472 | 3494644 | ||
| BC006789.1 | - | 4, 5 | 537 | 3589528 | ||

Ehf
Genomic Map