siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 8 | 784 | ||
| GenBank mRNA | AF035527.1 | - | 7 | 782 | ||
| AK028651.1 | - | 8 | 755 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehf| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 8 | 784 | ||
| GenBank mRNA | AF035527.1 | - | 7 | 782 | ||
| AK028651.1 | - | 8 | 755 | |||