siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010124.2 | NP_034254.1 | 1, 2 | 269 | ||
| GenBank mRNA | AK133912.1 | - | 1, 2 | 157 | ||
| AK148374.1 | - | 308 | ||||
| BC015082.1 | - | 2, 3 | 250 | 4218820 | ||
| BC119539.2 | - | 2, 3 | 233 | 40056467 | ||
| U75530.1 | - | 1, 2 | 266 | |||

Eif4ebp2
Genomic Map