siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010124.2 | NP_034254.1 | 2 | 404 | ||
| GenBank mRNA | AK133912.1 | - | 2 | 292 | ||
| AK148374.1 | - | 443 | ||||
| AK210066.1 | - | 1 | 142 | |||
| AK216468.1 | - | 1 | 142 | |||
| AK217241.1 | - | 1 | 142 | |||
| BC015082.1 | - | 3 | 385 | 4218820 | ||
| BC119539.2 | - | 3 | 368 | 40056467 | ||
| U75530.1 | - | 2 | 401 | |||

Eif4ebp2
Genomic Map