siRNA Details
| Gene Symbol: Aimp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007926.2 | NP_031952.2 | 6, 7 | 885 | ||
| GenBank mRNA | AK087932.1 | - | 6, 7 | 840 | ||
| AK133859.1 | - | 6, 7 | 838 | |||
| BC002054.1 | - | 6, 7 | 840 | 3496254 | ||
| BY631162.1 | - | 1, 2 | 172 | |||
| U10118.1 | - | 6, 7 | 836 | |||

Aimp1
Genomic Map