siRNA Details
| Gene Symbol: Emp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146346.1 | NP_001139818.1 | 3 | 417 | ||
| NM_010129.2 | NP_034259.2 | 3 | 388 | |||
| GenBank mRNA | AF011750.1 | - | 3 | 249 | ||
| AK146939.1 | - | 3 | 420 | |||
| BC001999.1 | - | 3 | 233 | 3485948 | ||
| CK022835.1 | - | 3 | 387 | |||
| U87948.1 | - | 3 | 366 | |||

Emp3
Genomic Map