siRNA Details
| Gene Symbol: Epb4.1l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013512.2 | NP_038540.2 | 22 | 2579 | ||
| GenBank mRNA | AK140545.1 | - | 3 | 718 | ||
| BC007166.1 | - | 22 | 2303 | 3490061 | ||
| BC011099.1 | - | 21 | 1873 | |||
| BC013557.1 | - | 22 | 2303 | 3490326 | ||
| BC089158.1 | - | 21 | 2148 | |||
| BE631863.1 | - | 2 | 146 | |||
| CX204315.1 | - | 9 | 688 | |||
| D28818.1 | - | 21 | 1847 | |||

Epb4.1l4a
Genomic Map