siRNA Details
| Gene Symbol: Epn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252188.1 | NP_001239117.1 | 3 | 532 | ||
| NM_001252189.1 | NP_001239118.1 | 2 | 459 | |||
| NM_010148.3 | NP_034278.1 | 2 | 459 | |||
| GenBank mRNA | AK036331.1 | - | 2 | 433 | ||
| AK139234.1 | - | 3 | 520 | |||
| AK139793.1 | - | 2 | 417 | |||
| AK160782.1 | - | 2 | 460 | |||
| AK173093.1 | - | 3 | 508 | |||
| BC039138.1 | - | 2 | 423 | 2647379 | ||

Epn2
Genomic Map