siRNA Details
Gene Symbol: Smarcad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007958.1 | NP_031984.1 | 4 | 864 | ||
GenBank mRNA | AK122454.1 | - | 4 | 671 | ||
AK134442.1 | - | 4 | 715 | |||
AK147884.1 | - | 4 | 653 | |||
X69942.1 | - | 4 | 864 |
