siRNA Details
| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007959.2 | NP_031985.2 | 5 | 802 | ||
| GenBank mRNA | AB221612.1 | - | 5 | 785 | ||
| BX638426.1 | - | 2 | 245 | |||
| L10427.1 | - | 5 | 802 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etv2| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007959.2 | NP_031985.2 | 5 | 802 | ||
| GenBank mRNA | AB221612.1 | - | 5 | 785 | ||
| BX638426.1 | - | 2 | 245 | |||
| L10427.1 | - | 5 | 802 | |||