siRNA Details
| Gene Symbol: Ewsr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001283061.1 | NP_001269990.1 | 4 | 210 | ||
| NM_001283062.1 | NP_001269991.1 | 4 | 210 | |||
| NM_001283063.1 | NP_001269992.1 | 4 | 210 | |||
| NM_007968.3 | NP_031994.2 | 4 | 210 | |||
| GenBank mRNA | AK019460.1 | - | 4 | 159 | ||
| AK034755.1 | - | 4 | 186 | |||
| AK046434.1 | - | 4 | 219 | |||
| AK049743.1 | - | 4 | 190 | |||
| AK146791.1 | - | 4 | 186 | |||
| AK147103.1 | - | 4 | 177 | |||
| AK147909.1 | - | 4 | 167 | |||
| AK150345.1 | - | 4 | 168 | |||
| AK151625.1 | - | 4 | 181 | |||
| AK153276.1 | - | 4 | 168 | |||
| BC068226.1 | - | 4 | 177 | 5707017 | ||
| BY272748.1 | - | 4 | 211 | |||
| X79233.1 | - | 4 | 199 | |||

Ewsr1
Genomic Map