siRNA Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080463.3 | NP_536711.3 | 5 | 770 | ||
| GenBank mRNA | AF375885.1 | - | 5 | 739 | ||
| AK044629.1 | - | 5 | 748 | |||
| AK053648.1 | - | 1 | 2276 | |||
| AK081059.1 | - | 5 | 727 | |||
| AK132301.1 | - | 5 | 725 | |||
| AK135635.1 | - | 5 | 752 | |||
| AK170038.1 | - | 5 | 725 | |||
| AK220326.1 | - | 1 | 2195 | |||
| BC046295.1 | - | 5 | 770 | 6485896 | ||

Pofut1
Genomic Map