siRNA Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080463.3 | NP_536711.3 | 6 | 1005 | ||
| GenBank mRNA | AF375885.1 | - | 6 | 974 | ||
| AK044629.1 | - | 6 | 983 | |||
| AK053648.1 | - | 2 | 2508 | |||
| AK081059.1 | - | 6 | 962 | |||
| AK132301.1 | - | 6 | 960 | |||
| AK135635.1 | - | 6 | 987 | |||
| AK220326.1 | - | 2 | 2430 | |||
| BC046295.1 | - | 6 | 1005 | 6485896 | ||

Pofut1
Genomic Map