siRNA Details
| Gene Symbol: Pofut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080463.3 | NP_536711.3 | 7 | 1083 | ||
| GenBank mRNA | AF375885.1 | - | 7 | 1052 | ||
| AK044629.1 | - | 7 | 1061 | |||
| AK053648.1 | - | 3 | 2586 | |||
| AK081059.1 | - | 7 | 1040 | |||
| AK132301.1 | - | 7 | 1038 | |||
| AK220326.1 | - | 3 | 2508 | |||
| BC046295.1 | - | 7 | 1083 | 6485896 | ||

Pofut1
Genomic Map