siRNA Details
| Gene Symbol: Acap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207223.1 | NP_997106.1 | 20 | 2372 | ||
| GenBank mRNA | AK147505.1 | - | 20 | 2426 | ||
| AK159055.1 | - | 16 | 2242 | |||
| BC067016.1 | - | 20 | 2372 | 6827624 | ||
| BC098196.1 | - | 19 | 2185 | 6847993 | ||

Acap3
Genomic Map