siRNA Details
| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242368.1 | NP_001229297.1 | 6 | 915 | ||
| NM_007972.4 | NP_031998.3 | 6 | 725 | |||
| GenBank mRNA | AF087644.1 | - | 5 | 462 | ||
| AJ222677.1 | - | 5 | 488 | |||
| AK149449.1 | - | 5 | 509 | |||
| AK154100.1 | - | 6 | 725 | |||
| AK154572.1 | - | 6 | 842 | |||
| AK170282.1 | - | 6 | 818 | |||
| AK171096.1 | - | 6 | 725 | |||
| BC003877.1 | - | 6 | 856 | 3495496 | ||
| BC050219.1 | - | 6 | 901 | 30018889 | ||
| CT010325.1 | - | 5 | 462 | |||

F10
Genomic Map