siRNA Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145400.3 | NP_663375.3 | 15 | 2466 | ||
| GenBank mRNA | AK028721.1 | - | 14 | 2137 | ||
| AK172893.1 | - | 15 | 2426 | |||
| BC006649.1 | - | 16 | 2605 | 3498834 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube4a| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145400.3 | NP_663375.3 | 15 | 2466 | ||
| GenBank mRNA | AK028721.1 | - | 14 | 2137 | ||
| AK172893.1 | - | 15 | 2426 | |||
| BC006649.1 | - | 16 | 2605 | 3498834 | ||