siRNA Details
| Gene Symbol: F2rl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007975.3 | NP_032001.2 | 2 | 637 | ||
| GenBank mRNA | AF080215.1 | - | 2 | 614 | ||
| AK036427.1 | - | 2 | 638 | |||
| AK153720.1 | - | 2 | 631 | |||
| AK156729.1 | - | 2 | 638 | |||
| BC119461.2 | - | 1 | 877 | 40045612 | ||

F2rl3
Genomic Map