siRNA Details
| Gene Symbol: F8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007978.3 | NP_032004.2 | 1 | 828 | ||
| GenBank mRNA | AF299331.1 | - | 1 | 857 | ||
| AK033184.1 | - | 1 | 830 | |||
| AK046025.1 | - | 1 | 844 | |||
| AK046459.1 | - | 1 | 843 | |||
| AK054140.1 | - | 1 | 829 | |||
| BC131938.1 | - | 1 | 856 | 40130215 | ||
| BC131940.1 | - | 1 | 856 | 40130217 | ||
| M83118.1 | - | 1 | 1109 | |||

F8a
Genomic Map