siRNA Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163776.1 | NP_001157248.1 | 6 | 663 | ||
| NM_080727.2 | NP_542765.2 | 5 | 523 | |||
| GenBank mRNA | AJ300738.1 | - | 5 | 523 | ||
| AK158203.1 | - | 6 | 666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss3| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163776.1 | NP_001157248.1 | 6 | 663 | ||
| NM_080727.2 | NP_542765.2 | 5 | 523 | |||
| GenBank mRNA | AJ300738.1 | - | 5 | 523 | ||
| AK158203.1 | - | 6 | 666 | |||