siRNA Details
| Gene Symbol: Fabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017399.4 | NP_059095.1 | 2 | 246 | ||
| GenBank mRNA | AK131868.1 | - | 2 | 256 | ||
| BC009812.1 | - | 2 | 246 | 4159971 | ||
| Y14660.1 | - | 2 | 246 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fabp1| Gene Symbol: Fabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017399.4 | NP_059095.1 | 2 | 246 | ||
| GenBank mRNA | AK131868.1 | - | 2 | 256 | ||
| BC009812.1 | - | 2 | 246 | 4159971 | ||
| Y14660.1 | - | 2 | 246 | |||