siRNA Details
| Gene Symbol: Ttbk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024856.2 | NP_001020027.1 | 15 | 2650 | ||
| NM_001024857.2 | NP_001020028.1 | 14 | 2576 | |||
| NM_080788.3 | NP_542966.2 | 15 | 2404 | |||
| GenBank mRNA | AK046978.1 | - | 15 | 2623 | ||
| AK077011.1 | - | 15 | 2426 | |||
| AK137024.1 | - | 14 | 2565 | |||
| AK148269.1 | - | 14 | 2580 | |||
| AK161858.1 | - | 2 | 668 | |||

Ttbk2
Genomic Map