siRNA Details
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016925.3 | NP_058621.2 | 36 | 3615 | ||
GenBank mRNA | AF178934.1 | - | 36 | 3611 | ||
AF208116.1 | - | 36 | 3617 | |||
AF208120.1 | - | 15 | 1698 | |||
BC150705.1 | - | 36 | 3613 | 9087616 |
