siRNA Details
| Gene Symbol: Fbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010180.2 | NP_034310.2 | 14 | 1783 | ||
| GenBank mRNA | AK035388.1 | - | 14 | 1736 | ||
| AK083573.1 | - | 14 | 1786 | |||
| AK085989.1 | - | 14 | 1726 | |||
| AK086451.1 | - | 14 | 1784 | |||
| AK137511.1 | - | 14 | 1730 | |||
| AK159646.1 | - | 14 | 1738 | |||
| AK159816.1 | - | 3 | 228 | |||
| BC007140.1 | - | 14 | 1727 | 3495754 | ||
| EU543224.1 | - | 14 | 1721 | |||
| X70854.1 | - | 14 | 1780 | |||

Fbln1
Genomic Map