siRNA Details
| Gene Symbol: Fgf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010197.3 | NP_034327.1 | 4 | 480 | ||
| GenBank mRNA | AK035330.1 | - | 4 | 481 | ||
| AK142163.1 | - | 5 | 557 | |||
| BC027001.1 | - | 4 | 469 | |||
| BC037601.1 | - | 4 | 499 | 5137246 | ||
| M30641.1 | - | 3 | 293 | |||
| U67610.1 | - | 4 | 446 | |||

Fgf1
Genomic Map