siRNA Details
| Gene Symbol: Fgf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008003.2 | NP_032029.1 | 1 | 329 | ||
| GenBank mRNA | AF007268.1 | - | 1 | 329 | ||
| AK017829.1 | - | 1 | 331 | |||
| AK052093.1 | - | 1 | 331 | |||
| BC021328.1 | - | 1 | 321 | 5066286 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK050685.1 | - | 429 | Chr. - - [-] on Build | ||

Fgf15
Genomic Map